This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01177075_m1
See other BCKDHB GE Assays ›
Gene Symbol:
Bckdhb
Gene Name:
branched chain ketoacid dehydrogenase E1, beta polypeptide
Gene Aliases:
BCKDE1B, BCKDH E1-beta
Chromosome Location:
Chr.9: 83925111 - 84124240 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01177075_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Bckdhb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_199195.1 NP_954665.1 5 - 6 573 145
GenBank mRNA AK089921.1 - 5 - 6 653 147
AK156264.1 - 5 - 6 652 145
BC064099.1 - 5 - 6 573 1246285 145
L16992.1 - 5 - 6 645 145

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900001103.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Bckdhb

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

metabolic process
cellular amino acid catabolic process
branched-chain amino acid catabolic process
oxidation-reduction process
catalytic activity
alpha-ketoacid dehydrogenase activity
3-methyl-2-oxobutanoate dehydrogenase (2-methylpropanoyl-transferring) activity
oxidoreductase activity
protein complex binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900008687.mm.1 NM_199195.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900008687.mm.1 NM_199195.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900008687.mm.1 TC0900001103.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900008688.mm.1 NM_199195.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900008688.mm.1 NM_199195.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900008688.mm.1 TC0900001103.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0