This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01176766_m1
See other STT3A GE Assays ›
Gene Symbol:
Stt3a
Gene Name:
STT3, subunit of the oligosaccharyltransferase complex, homolog A (S. cerevisiae)
Gene Aliases:
AA408947, BB081708, Itm1
Chromosome Location:
Chr.9: 36729345 - 36767674 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01176766_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Stt3a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008408.4 NP_032434.3 13 - 14 1626 69
GenBank mRNA AK030363.1 - 13 - 14 1690 69
AK077877.1 - 13 - 14 1650 69
AK081547.1 - 13 - 14 1683 69
AK087470.1 - 13 - 14 1672 69
AK129027.1 - 13 - 14 1664 69
AK150640.1 - 13 - 14 1671 69
AK151541.1 - 13 - 14 1682 69
AK151791.1 - 13 - 14 1682 69
AK152242.1 - 13 - 14 1671 69
AK153840.1 - 13 - 14 1627 69
AK153886.1 - 13 - 14 1727 69
AK160037.1 - 13 - 14 1665 69
AK160607.1 - 13 - 14 1627 69
AK168505.1 - 13 - 14 1672 69
AK168937.1 - 13 - 14 1671 69
AK169669.1 - 13 - 14 1650 69
AK213353.1 - 1 - 2 144 69
AX799090.1 - 13 - 14 1669 69
BC028897.1 - 12 - 13 1575 69
BC037612.1 - 13 - 14 1665 5353797 69
BC085313.1 - 13 - 14 1655 30443880 69
L34260.1 - 13 - 14 1669 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900002037.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Stt3a

Panther Classification:

Molecular Function -

glycosyltransferase

Gene Ontology Categories:

Function(s) Process(es)

protein glycosylation
protein N-linked glycosylation via asparagine
co-translational protein modification
oligosaccharyl transferase activity
dolichyl-diphosphooligosaccharide-protein glycotransferase activity
transferase activity
transferase activity, transferring glycosyl groups

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900016928.mm.1 NM_008408.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016928.mm.1 NM_008408.4 Refseq
Affymetrix Mouse Clariom S Array PSR0900016928.mm.1 TC0900002037.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0