This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01176410_g1
See other TBRG1 GE Assays ›
Gene Symbol:
Tbrg1
Gene Name:
transforming growth factor beta regulated gene 1
Gene Aliases:
AA408552, AA409675, Niam, Tb-5, mFLJ00213
Chromosome Location:
Chr.9: 37649182 - 37657312 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01176410_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tbrg1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025289.3 NP_079565.2 4 - 5 740 74
GenBank mRNA AK011268.1 - 4 - 5 664 74
AK150549.1 - 4 - 5 667 74
AK151209.1 - 4 - 5 634 74
AK151457.1 - 4 - 5 652 74
AK166547.1 - 4 - 5 662 74
AK218348.1 - 1 - 2 46 74
BC021331.1 - 4 - 5 751 5003215 74
BC040813.1 - 4 - 5 653 4223194 74
BC043022.1 - 4 - 5 655 5688749 74
BC065795.1 - 4 - 5 653 1529903 74
DQ144541.1 - 4 - 5 572 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900002054.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tbrg1

Panther Classification:

Molecular Function -

growth factor intercellular signal molecule

Gene Ontology Categories:

Function(s) Process(es)

DNA replication
cell cycle
cell cycle arrest
negative regulation of cell proliferation
nucleolus to nucleoplasm transport
protein stabilization
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900017142.mm.1 NM_025289.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900017142.mm.1 NM_025289.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900017142.mm.1 TC0900002054.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0