This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01175850_m1
See other TYK2 GE Assays ›
Gene Symbol:
Tyk2
Gene Name:
tyrosine kinase 2
Gene Aliases:
JTK1
Chromosome Location:
Chr.9: 21104068 - 21131275 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01175850_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tyk2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001205312.1 NP_001192241.1 7 - 8 1339 67
NM_018793.2 NP_061263.2 7 - 8 1270 67
GenBank mRNA AF173032.1 - 7 - 8 1205 67
AK154438.1 - 7 - 8 1297 67
AK155923.1 - 7 - 8 1183 67
AK159303.1 - 7 - 8 1239 67
AK167726.1 - 7 - 8 1249 67
AK170107.1 - 7 - 8 1209 67
AK170940.1 - 7 - 8 1209 67
AK171609.1 - 7 - 8 1183 67
BC094240.1 - 6 - 7 1166 30610201 67

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tyk2

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
transmembrane receptor protein tyrosine kinase signaling pathway
phosphorylation
cell migration
peptidyl-tyrosine phosphorylation
cell differentiation
intracellular signal transduction
peptidyl-tyrosine autophosphorylation
regulation of cell proliferation
innate immune response
nucleotide binding
protein kinase activity
protein tyrosine kinase activity
non-membrane spanning protein tyrosine kinase activity
growth hormone receptor binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
type 1 angiotensin receptor binding
hormone receptor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0