This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01175805_m1
See other BMPER GE Assays ›
Gene Symbol:
Bmper
Gene Name:
BMP-binding endothelial regulator
Gene Aliases:
3110056H04Rik, CV-2, Crim3, Cv2
Chromosome Location:
Chr.9: 23223076 - 23485215 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01175805_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Bmper
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028472.2 NP_082748.1 13 - 14 2306 108
GenBank mRNA AF454954.1 - 13 - 14 2306 108
AK014221.1 - 11 - 12 1511 108
AY263358.1 - 13 - 14 1827 108
BC042718.1 - 1 - 2 316 108
BC066153.1 - 13 - 14 1811 30098453 108

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900000226.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Bmper

Panther Classification:

Molecular Function -

extracellular matrix protein

Gene Ontology Categories:

Function(s) Process(es)

blood vessel endothelial cell proliferation involved in sprouting angiogenesis
regulation of endothelial cell migration
negative regulation of BMP signaling pathway
endothelial cell activation
endothelial cell differentiation
inner ear development
regulation of pathway-restricted SMAD protein phosphorylation
positive regulation of ERK1 and ERK2 cascade
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900001756.mm.1 NM_028472.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900001756.mm.1 TC0900000226.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0