This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01175786_g1
See other GM30400 GE Assays ›
Gene Symbol:
Gm30400
Gene Name:
predicted gene, 30400
Gene Aliases:
-
Chromosome Location:
Chr.10: 78747130 - 78767585 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01175786_g1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gm30400
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK079520.1 - 2 - 3 1445 81
AK164315.1 - 4 - 5 661 81

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream coiled-coil domain containing 105 Ccdc105 4931413A09Rik -11.2kb Anti-sense Chr. 10 - 78746924 - 78753067 on NCBI Build GRCm38
Downstream solute carrier family 1 (high affinity aspartate/glutamate transporter),
member 6
Slc1a6 EAAT4 13.3kb Sense Chr. 10 - 78780496 - 78814825 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0