Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900021122.mm.1 |
NM_019797.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900021122.mm.1 |
NM_019797.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900021122.mm.1 |
TC0900002519.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900021124.mm.1 |
NM_019797.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900021124.mm.1 |
NM_019797.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900021124.mm.1 |
TC0900002519.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900021123.mm.1 |
NM_019797.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900021123.mm.1 |
NM_019797.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900021123.mm.1 |
TC0900002519.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900021119.mm.1 |
NM_019797.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900021119.mm.1 |
NM_019797.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900021119.mm.1 |
TC0900002519.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900021125.mm.1 |
NM_019797.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900021125.mm.1 |
NM_019797.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900021125.mm.1 |
TC0900002519.mm.1 |
MTA TC |