This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01172982_m1
See other HNRNPL GE Assays ›
Gene Symbol:
Hnrnpl
Gene Name:
heterogeneous nuclear ribonucleoprotein L
Gene Aliases:
C79783, D830027H13Rik, Hnrpl
Chromosome Location:
Chr.7: 28808546 - 28822268 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01172982_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hnrnpl
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_177301.5 NP_796275.3 6 - 7 896 62
GenBank mRNA AK144684.1 - 6 - 7 850 62
BC027206.1 - 6 - 7 873 4218471 62
BC099683.1 - 6 - 7 891 6530863 62

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700000513.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hnrnpl

Gene Ontology Categories:

Function(s) Process(es)

mRNA processing
cellular response to amino acid starvation
positive regulation of translation
negative regulation of mRNA splicing, via spliceosome
positive regulation of mRNA binding
nucleotide binding
nucleic acid binding
RNA binding
mRNA 3'-UTR binding
transcription regulatory region DNA binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700004244.mm.1 NM_177301.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700004244.mm.1 NM_177301.5 Refseq
Affymetrix Mouse Clariom S Array PSR0700004244.mm.1 TC0700000513.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-fd98957d6-rbs6c:80/100.66.128.142:80.
git-commit: 278a916a254c4ca07f9ccbb2b9efa8cd7578b5be
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0