This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01172882_g1
See other MATK GE Assays ›
Gene Symbol:
Matk
Gene Name:
megakaryocyte-associated tyrosine kinase
Gene Aliases:
CHK, HYL, Ntk
Chromosome Location:
Chr.10: 81252795 - 81262982 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01172882_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Matk
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010768.2 NP_034898.1 1 - 2 281 119

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_010768.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Matk

Gene Ontology Categories:

Function(s) Process(es)

morphogenesis of an epithelium
protein phosphorylation
transmembrane receptor protein tyrosine kinase signaling pathway
phosphorylation
cell migration
cell differentiation
peptidyl-tyrosine autophosphorylation
regulation of cell proliferation
innate immune response
nucleotide binding
protein kinase activity
protein tyrosine kinase activity
non-membrane spanning protein tyrosine kinase activity
receptor binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000006942.mm.1 NM_010768.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000006942.mm.1 TC1000000889.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000006943.mm.1 NM_010768.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000006943.mm.1 TC1000000889.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0