This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01172872_g1
See other MFSD12 GE Assays ›
Gene Symbol:
Mfsd12
Gene Name:
major facilitator superfamily domain containing 12
Gene Aliases:
C19orf28, F630110N24Rik, Wdt1
Chromosome Location:
Chr.10: 81357469 - 81366225 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01172872_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mfsd12
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028657.3 NP_082933.2 7 - 8 1206 80
GenBank mRNA AK149678.1 - 7 - 8 1212 80
AK154386.1 - 7 - 8 1208 80
AK170185.1 - 7 - 8 1225 80
AK171084.1 - 7 - 8 1208 80
BC027706.1 - 1 - 2 154 80
BC038699.1 - 1 - 2 154 80
BC118620.1 - 7 - 8 1206 40107417 80
BC119790.1 - 7 - 8 1206 40107416 80

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000000895.mm.1

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

transport
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000007066.mm.1 NM_028657.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000007066.mm.1 NM_028657.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000007066.mm.1 TC1000000895.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0