Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1600001913.mm.1 |
NM_001164609.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600001913.mm.1 |
NM_001164609.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600001913.mm.1 |
NM_001164610.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600001913.mm.1 |
NM_001164610.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600001913.mm.1 |
NM_001164611.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600001913.mm.1 |
NM_001164611.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600001913.mm.1 |
NM_029945.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600001913.mm.1 |
NM_029945.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600001913.mm.1 |
TC1600000221.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600001909.mm.1 |
NM_001164609.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600001909.mm.1 |
NM_001164609.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600001909.mm.1 |
NM_001164610.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600001909.mm.1 |
NM_001164610.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600001909.mm.1 |
NM_001164611.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600001909.mm.1 |
NM_001164611.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600001909.mm.1 |
NM_029945.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600001909.mm.1 |
NM_029945.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600001909.mm.1 |
TC1600000221.mm.1 |
MTA TC |