This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01171186_m1
See other SMPD4 GE Assays ›
Gene Symbol:
Smpd4
Gene Name:
sphingomyelin phosphodiesterase 4
Gene Aliases:
4122402O22Rik, mKIAA1418
Chromosome Location:
Chr.16: 17619354 - 17644830 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01171186_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Smpd4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_029945.3 NP_084221.2 11 - 12 1070 114
GenBank mRNA AK019466.2 - 11 - 12 1071 114
AK129354.1 - 11 - 12 1047 114

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600000221.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Smpd4

Panther Classification:

Molecular Function -

hydrolase phosphodiesterase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

sphingomyelin catabolic process
glycerophospholipid catabolic process
ceramide biosynthetic process
cellular response to tumor necrosis factor
sphingomyelin phosphodiesterase activity
hydrolase activity
metal ion binding
sphingomyelin phosphodiesterase D activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600001942.mm.1 NM_001164609.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600001942.mm.1 NM_001164609.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600001942.mm.1 NM_001164610.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600001942.mm.1 NM_001164610.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600001942.mm.1 NM_001164611.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600001942.mm.1 NM_001164611.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600001942.mm.1 NM_029945.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600001942.mm.1 NM_029945.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600001942.mm.1 TC1600000221.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600001941.mm.1 NM_001164609.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600001941.mm.1 NM_001164609.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600001941.mm.1 NM_001164610.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600001941.mm.1 NM_001164610.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600001941.mm.1 NM_001164611.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600001941.mm.1 NM_001164611.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600001941.mm.1 NM_029945.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600001941.mm.1 NM_029945.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600001941.mm.1 TC1600000221.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600001938.mm.1 NM_029945.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600001938.mm.1 NM_029945.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600001938.mm.1 TC1600000221.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600001940.mm.1 NM_001164609.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600001940.mm.1 NM_001164609.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600001940.mm.1 NM_001164610.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600001940.mm.1 NM_001164610.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600001940.mm.1 NM_001164611.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600001940.mm.1 NM_001164611.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600001940.mm.1 NM_029945.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600001940.mm.1 NM_029945.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600001940.mm.1 TC1600000221.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0