This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01170334_m1
See other ANLN GE Assays ›
Gene Symbol:
Anln
Gene Name:
anillin, actin binding protein
Gene Aliases:
1110037A17Rik, 2900037I21Rik, C78101, Scraps
Chromosome Location:
Chr.9: 22331214 - 22389696 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01170334_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Anln
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028390.3 NP_082666.2 9 - 10 1832 68
GenBank mRNA AK045703.1 - 6 - 7 796 68
BC030489.1 - 2 - 3 174 5373071 68
BC032164.1 - 9 - 10 1850 5346703 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900001908.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Anln

Panther Classification:

Molecular Function -

non-motor actin binding protein

Gene Ontology Categories:

Function(s) Process(es)

mitotic cytokinesis
hematopoietic progenitor cell differentiation
cell cycle
mitotic nuclear division
cell division
glomerular visceral epithelial cell migration
actin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900016083.mm.1 NM_028390.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016083.mm.1 NM_028390.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900016083.mm.1 TC0900001908.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0