This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01169262_m1
See other SIPA1L3 GE Assays ›
Gene Symbol:
Sipa1l3
Gene Name:
signal-induced proliferation-associated 1 like 3
Gene Aliases:
2610511M17Rik
Chromosome Location:
Chr.7: 29320372 - 29518943 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01169262_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sipa1l3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081028.1 NP_001074497.1 7 - 8 2634 113
GenBank mRNA AK154121.1 - 7 - 8 2636 114
AK163313.1 - 4 - 5 1016 113
AK171182.1 - 6 - 7 2558 113

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700002723.mm.1

Set Membership:

Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

hematopoietic progenitor cell differentiation
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700024114.mm.1 NM_001081028.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700024114.mm.1 NM_001081028.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700024114.mm.1 TC0700002723.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0