This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01169074_m1
See other SUPV3L1 GE Assays ›
Gene Symbol:
Supv3l1
Gene Name:
suppressor of var1, 3-like 1 (S. cerevisiae)
Gene Aliases:
6330443E10Rik
Chromosome Location:
Chr.10: 62429329 - 62451294 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01169074_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Supv3l1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_181423.2 NP_852088.1 4 - 5 580 143
GenBank mRNA AK031911.1 - 4 - 5 571 143
AK143755.1 - 4 - 5 605 143
BC049796.1 - 4 - 5 580 6332552 143

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000002258.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Supv3l1

Panther Classification:

Molecular Function -

DNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

mitochondrial mRNA catabolic process
positive regulation of mitochondrial RNA catabolic process
mitochondrial RNA 3'-end processing
DNA recombination
RNA catabolic process
positive regulation of cell growth
DNA duplex unwinding
mitochondrial ncRNA surveillance
mitochondrial mRNA surveillance
negative regulation of apoptotic process
mitochondrion morphogenesis
chromatin maintenance
mitochondrial RNA surveillance
nucleotide binding
DNA binding
DNA helicase activity
double-stranded RNA binding
ATP-dependent RNA helicase activity
helicase activity
ATP binding
hydrolase activity
hydrolase activity, acting on acid anhydrides
3'-5' RNA helicase activity
protein homodimerization activity
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000016099.mm.1 NM_181423.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000016099.mm.1 NM_181423.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000016099.mm.1 TC1000002258.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0