This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01167285_m1
See other DNAJC2 GE Assays ›
Gene Symbol:
Dnajc2
Gene Name:
DnaJ (Hsp40) homolog, subfamily C, member 2
Gene Aliases:
AU020218, MIDA1, Zrf1, Zrf2
Chromosome Location:
Chr.5: 21757264 - 21785248 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01167285_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dnajc2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009584.4 NP_033610.1 1 - 2 175 63
GenBank mRNA AK013295.1 - 1 - 2 166 63
AK131964.1 - 1 - 2 196 63
AK136951.1 - 1 - 2 199 63
AK161347.1 - 1 - 2 143 63
AK161376.1 - 1 - 2 196 63
AK162409.1 - 1 - 2 196 63
AK169850.1 - 1 - 2 210 63
BC021755.1 - 1 - 2 180 63
BC052027.1 - 1 - 2 202 5718637 63
D63784.1 - 1 - 2 174 63
U53208.1 - 1 - 2 130 63

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK161347.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dnajc2

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

DNA replication
transcription, DNA-templated
regulation of transcription, DNA-templated
chromatin modification
negative regulation of cell growth
positive regulation of transcription, DNA-templated
negative regulation of DNA biosynthetic process
DNA binding
chromatin binding
protein binding
Hsp70 protein binding
histone binding
ubiquitin binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500018410.mm.1 NM_009584.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500018410.mm.1 NM_009584.4 Refseq
Affymetrix Mouse Clariom S Array PSR0500018410.mm.1 TC0500002025.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 656913 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0