This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01167283_g1
See other DNAJC2 GE Assays ›
Gene Symbol:
Dnajc2
Gene Name:
DnaJ (Hsp40) homolog, subfamily C, member 2
Gene Aliases:
AU020218, MIDA1, Zrf1, Zrf2
Chromosome Location:
Chr.5: 21757264 - 21785248 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01167283_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dnajc2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009584.4 NP_033610.1 14 - 15 1638 128
GenBank mRNA AK131964.1 - 14 - 15 1659 128
AK167991.1 - 14 - 15 1659 128
BC021755.1 - 13 - 14 3383 128
BC052027.1 - 14 - 15 1665 5718637 128
D63784.1 - 14 - 15 1637 128

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dnajc2

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

DNA replication
transcription, DNA-templated
regulation of transcription, DNA-templated
chromatin modification
negative regulation of cell growth
positive regulation of transcription, DNA-templated
negative regulation of DNA biosynthetic process
DNA binding
chromatin binding
protein binding
Hsp70 protein binding
histone binding
ubiquitin binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500018388.mm.1 NM_009584.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500018388.mm.1 NM_009584.4 Refseq
Affymetrix Mouse Clariom S Array PSR0500018388.mm.1 TC0500002025.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0