This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01167046_g1
See other GDPD4 GE Assays ›
Gene Symbol:
Gdpd4
Gene Name:
glycerophosphodiester phosphodiesterase domain containing 4
Gene Aliases:
4921513O04
Chromosome Location:
Chr.7: 97919944 - 98049663 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01167046_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gdpd4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_177696.3 NP_808364.2 9 - 10 964 100
GenBank mRNA AK029527.1 - 9 - 10 968 100
AK161500.1 - 9 - 10 931 100
BC140990.1 - 7 - 8 692 9055602 100

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700001363.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gdpd4

Panther Classification:

Molecular Function -

phosphodiesterase

Gene Ontology Categories:

Function(s) Process(es)

glycerol metabolic process
lipid metabolic process
biological_process
molecular_function
nuclease activity
phosphoric diester hydrolase activity
glycerophosphodiester phosphodiesterase activity
hydrolase activity
phosphoric ester hydrolase activity
T/G mismatch-specific endonuclease activity
retroviral 3' processing activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700011594.mm.1 NM_177696.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700011594.mm.1 NM_177696.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700011594.mm.1 TC0700001363.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700011593.mm.1 NM_177696.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700011593.mm.1 NM_177696.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700011593.mm.1 TC0700001363.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0