This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01166883_g1
See other PDK4 GE Assays ›
Gene Symbol:
Pdk4
Gene Name:
pyruvate dehydrogenase kinase, isoenzyme 4
Gene Aliases:
AV005916
Chromosome Location:
Chr.6: 5483351 - 5496278 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01166883_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pdk4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013743.2 NP_038771.1 8 - 9 984 86
GenBank mRNA AJ001418.1 - 8 - 9 984 86
AK004543.1 - 8 - 9 1009 86
AK036818.1 - 8 - 9 1016 86
BC026134.1 - 8 - 9 1013 4188580 86

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600001810.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pdk4

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of defense response to virus by host
carbohydrate metabolic process
glucose metabolic process
acetyl-CoA biosynthetic process from pyruvate
protein phosphorylation
regulation of pH
insulin receptor signaling pathway
cellular response to starvation
regulation of acetyl-CoA biosynthetic process from pyruvate
regulation of cellular ketone metabolic process
regulation of glucose metabolic process
phosphorylation
regulation of fatty acid biosynthetic process
glucose homeostasis
response to starvation
regulation of bone resorption
regulation of fatty acid oxidation
cellular response to fatty acid
reactive oxygen species metabolic process
negative regulation of anoikis
nucleotide binding
protein kinase activity
protein serine/threonine/tyrosine kinase activity
pyruvate dehydrogenase (acetyl-transferring) kinase activity
ATP binding
kinase activity
transferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600014356.mm.1 NM_013743.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600014356.mm.1 NM_013743.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600014356.mm.1 TC0600001810.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0