This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01166085_g1
See other ATF6B GE Assays ›
Gene Symbol:
Atf6b
Gene Name:
activating transcription factor 6 beta
Gene Aliases:
AA617266, ATF6beta, Creb-rp, Crebl1
Chromosome Location:
Chr.17: 34647146 - 34655074 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01166085_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atf6b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_017406.4 NP_059102.2 7 - 8 776 119
GenBank mRNA AK090378.1 - 7 - 8 723 119
AK143765.1 - 7 - 8 709 119
AK146671.1 - 7 - 8 743 119
AK159169.1 - 7 - 8 709 119
AK170027.1 - 7 - 8 709 119
AK179180.1 - 3 - 4 298 119
BC013534.1 - 7 - 8 776 4239106 119
BC052635.1 - 7 - 8 711 6395626 119

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Atf6b

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
response to unfolded protein
endoplasmic reticulum unfolded protein response
positive regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
DNA binding
transcription factor activity, sequence-specific DNA binding
cAMP response element binding
sequence-specific DNA binding
transcription regulatory region DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700006285.mm.1 NM_017406.4 Refseq
Affymetrix Mouse Clariom S Array PSR1700006285.mm.1 TC1700000643.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0