This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01165943_g1
See other METTL1 GE Assays ›
Gene Symbol:
Mettl1
Gene Name:
methyltransferase like 1
Gene Aliases:
2810012D02Rik
Chromosome Location:
Chr.10: 127041643 - 127046365 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01165943_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mettl1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010792.1 NP_034922.1 3 - 4 490 71
GenBank mRNA AB023619.1 - 3 - 4 490 71
AK140605.1 - 3 - 4 465 71
AK160562.1 - 3 - 4 469 71
AK160914.1 - 3 - 4 467 71
BC012649.1 - 3 - 4 499 4020487 71
Y18644.1 - 3 - 4 436 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000001544.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mettl1

Panther Classification:

Molecular Function -

RNA processing factor

Gene Ontology Categories:

Function(s) Process(es)

tRNA modification
tRNA processing
tRNA methylation
methylation
RNA (guanine-N7)-methylation
tRNA binding
RNA binding
methyltransferase activity
tRNA (guanine-N7-)-methyltransferase activity
transferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000010914.mm.1 NM_010792.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000010914.mm.1 NM_010792.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000010914.mm.1 TC1000001544.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0