This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01165796_m1
See other 4930430F21RIK GE Assays ›
Gene Symbol:
4930430F21Rik
Gene Name:
RIKEN cDNA 4930430F21 gene
Gene Aliases:
-
Chromosome Location:
Chr.15: 31039126 - 31043331 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01165796_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4930430F21Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_045186.1 3 - 4 477 79
GenBank mRNA AK015252.1 - 3 - 4 485 79

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream catenin (cadherin associated protein),
delta 2
Ctnnd2 Catnd2,
Nprap,
neurojugin
-11.8kb Sense Chr. 15 - 30172593 - 31029344 on NCBI Build GRCm38
Downstream death-associated protein Dap 4921531N22Rik,
AI789753,
AU040360
181.1kb Sense Chr. 15 - 31224385 - 31274338 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500000197.mm.1

Set Membership:

Amplicon length between 71 and 85 3' Most Mus musculus 1700 Probe spans exons

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 761134 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P106606 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P106606 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102184 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P106606 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30112117 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30112124 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12335585 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30112133 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30112091 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30112154 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30112086 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30112157 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30112082 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30112121 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30112151 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30112094 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12335593 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12197854 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30112128 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30112126 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30112098 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30112162 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30112167 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30112165 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12197891 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12335600 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30112087 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30112136 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30112160 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12197889 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30112159 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30112156 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30112089 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30112099 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30112131 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30112163 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30112142 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12197856 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30112093 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30112083 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12197895 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30112097 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12197890 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500001498.mm.1 NR_045186.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500001498.mm.1 NR_045186.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500001498.mm.1 TC1500000197.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500001499.mm.1 NR_045186.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500001499.mm.1 NR_045186.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500001499.mm.1 TC1500000197.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 340346 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0