This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01165070_m1
See other SLC16A7 GE Assays ›
Gene Symbol:
Slc16a7
Gene Name:
solute carrier family 16 (monocarboxylic acid transporters), member 7
Gene Aliases:
4921534N07Rik, Mct2
Chromosome Location:
Chr.10: 125219284 - 125389562 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01165070_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc16a7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011391.1 NP_035521.1 2 - 3 328 100
GenBank mRNA AF058054.1 - 2 - 3 328 100

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF058054.1, NM_011391.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Transporter Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc16a7

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
monocarboxylic acid transport
lactate transmembrane transport
plasma membrane lactate transport
transmembrane transport
pyruvate transmembrane transport
monocarboxylic acid transmembrane transporter activity
lactate transmembrane transporter activity
symporter activity
pyruvate transmembrane transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000022947.mm.1 NM_011391.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000022947.mm.1 NM_011391.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000022947.mm.1 TC1000003078.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000022948.mm.1 NM_011391.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000022948.mm.1 NM_011391.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000022948.mm.1 TC1000003078.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000022949.mm.1 NM_011391.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000022949.mm.1 NM_011391.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000022949.mm.1 TC1000003078.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000022952.mm.1 NM_011391.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000022952.mm.1 NM_011391.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000022952.mm.1 TC1000003078.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000022953.mm.1 NM_011391.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000022953.mm.1 NM_011391.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000022953.mm.1 TC1000003078.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0