This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01164863_g1
See other GLS2 GE Assays ›
Gene Symbol:
Gls2
Gene Name:
glutaminase 2 (liver, mitochondrial)
Gene Aliases:
A330074B06Rik, AI195532, GA, GLS, Lga
Chromosome Location:
Chr.10: 128194635 - 128210004 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01164863_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gls2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001033264.3 NP_001028436.2 2 - 3 372 78
NM_001285777.1 NP_001272706.1 2 - 3 372 78
NM_001285779.1 NP_001272708.1 2 - 3 372 78
GenBank mRNA AK039618.1 - 2 - 3 260 78
AK165896.1 - 2 - 3 318 78
AK220231.1 - 2 - 3 324 78
BC141115.1 - 2 - 3 372 9055727 78

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001285777.1, NM_001285779.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gls2

Panther Classification:

Molecular Function -

hydrolase

Gene Ontology Categories:

Function(s) Process(es)

glutamine metabolic process
regulation of apoptotic process
reactive oxygen species metabolic process
positive regulation of protein targeting to mitochondrion
molecular_function
glutaminase activity
hydrolase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0