This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01164237_g1
See other S100G GE Assays ›
Gene Symbol:
S100g
Gene Name:
S100 calcium binding protein G
Gene Aliases:
AU042539, CABP1, CaBP-D9K, Cabp9k, Calb3
Chromosome Location:
Chr.X: 162961992 - 162964599 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01164237_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: S100g
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009789.2 NP_033919.1 1 - 2 86 58
GenBank mRNA AF136283.1 - 1 - 2 49 58
AF293948.1 - 1 - 2 86 58
AF293949.1 - 1 - 2 86 58
AK146558.1 - 1 - 2 57 58
BC010751.1 - 1 - 2 35 4237050 58

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF136283.1, AF293948.1, AF293949.1, AK146558.1, BC010751.1, NM_009789.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene S100g

Panther Classification:

Molecular Function -

calmodulin-related

Gene Ontology Categories:

Process(es)

vitamin D binding
calcium ion binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00020665.mm.1 NM_009789.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00020665.mm.1 NM_009789.2 Refseq
Affymetrix Mouse Clariom S Array PSR0X00020665.mm.1 TC0X00003316.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00020663.mm.1 NM_009789.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00020663.mm.1 NM_009789.2 Refseq
Affymetrix Mouse Clariom S Array PSR0X00020663.mm.1 TC0X00003316.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00020664.mm.1 NM_009789.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00020664.mm.1 NM_009789.2 Refseq
Affymetrix Mouse Clariom S Array PSR0X00020664.mm.1 TC0X00003316.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0