This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01161295_g1
See other DXO GE Assays ›
Gene Symbol:
Dxo
Gene Name:
decapping exoribonuclease
Gene Aliases:
Dom3z, NG6
Chromosome Location:
Chr.17: 34837019 - 34839186 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01161295_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dxo
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163770.1 NP_001157242.1 1 - 2 197 103
GenBank mRNA CJ061424.1 - 1 - 2 198 103

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the NM_001163770.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

mRNA catabolic process
metabolic process
RNA destabilization
nuclear mRNA surveillance
nucleic acid phosphodiester bond hydrolysis
nucleotide binding
3'-5'-exoribonuclease activity
magnesium ion binding
RNA binding
mRNA binding
nuclease activity
exonuclease activity
5'-3' exoribonuclease activity
thiamine-pyrophosphatase activity
5'-3' exonuclease activity
8-oxo-7,8-dihydroguanosine triphosphate pyrophosphatase activity
UDP-2,3-diacylglucosamine hydrolase activity
bis(5'-nucleosyl)-tetraphosphatase activity
dATP pyrophosphohydrolase activity
pyrophosphatase activity
hydrolase activity
dihydroneopterin monophosphate phosphatase activity
dihydroneopterin triphosphate pyrophosphohydrolase activity
RNA pyrophosphohydrolase activity
dITP diphosphatase activity
dTTP diphosphatase activity
XTP diphosphatase activity
ATP-dependent 5'-3' DNA helicase activity
phosphocholine hydrolase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700006367.mm.1 NM_001163770.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700006367.mm.1 NM_001163770.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700006367.mm.1 NM_033613.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700006367.mm.1 NM_033613.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700006367.mm.1 TC1700000646.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700006365.mm.1 NM_001163770.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700006365.mm.1 NM_001163770.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700006365.mm.1 NM_033613.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700006365.mm.1 NM_033613.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700006365.mm.1 TC1700000646.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700006371.mm.1 NM_001163770.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700006371.mm.1 NM_001163770.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700006371.mm.1 NM_033613.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700006371.mm.1 NM_033613.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700006371.mm.1 TC1700000646.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0