This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01160350_m1
See other NAB2 GE Assays ›
Gene Symbol:
Nab2
Gene Name:
Ngfi-A binding protein 2
Gene Aliases:
AI451907
Chromosome Location:
Chr.10: 127660918 - 127666703 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01160350_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nab2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008668.2 NP_032694.2 6 - 7 1718 101
GenBank mRNA AK034709.1 - 1 - 2 194 101
AK171180.1 - 6 - 7 1598 101
BC045139.1 - 6 - 7 1602 6306757 101

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000003120.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nab2

Gene Ontology Categories:

Function(s) Process(es)

endochondral ossification
transcription, DNA-templated
regulation of transcription, DNA-templated
Schwann cell differentiation
negative regulation of transcription from RNA polymerase III promoter
myelination
regulation of epidermis development
negative regulation of transcription, DNA-templated
transcription factor binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000023434.mm.1 NM_001122895.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000023434.mm.1 NM_001122895.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000023434.mm.1 NM_008668.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000023434.mm.1 NM_008668.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000023434.mm.1 TC1000003120.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 689484 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header