This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01159333_g1
See other APOM GE Assays ›
Gene Symbol:
Apom
Gene Name:
apolipoprotein M
Gene Aliases:
1190010O19Rik, G3a, NG20
Chromosome Location:
Chr.17: 35128997 - 35131752 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01159333_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Apom
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_018816.1 NP_061286.1 1 - 2 143 77
GenBank mRNA AF207820.1 - 1 - 2 143 77
AK004530.1 - 1 - 2 188 77
BC021597.1 - 1 - 2 180 5100655 77

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Apom

Panther Classification:

Molecular Function -

apolipoprotein transfer/carrier protein

Gene Ontology Categories:

Function(s) Process(es)

transport
lipid transport
cholesterol efflux
high-density lipoprotein particle remodeling
high-density lipoprotein particle assembly
high-density lipoprotein particle clearance
negative regulation of plasma lipoprotein particle oxidation
lipoprotein metabolic process
reverse cholesterol transport
lipid transporter activity
phospholipid binding
antioxidant activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700018515.mm.1 NM_018816.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700018515.mm.1 NM_018816.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700018515.mm.1 TC1700001949.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700018516.mm.1 NM_018816.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700018516.mm.1 NM_018816.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700018516.mm.1 TC1700001949.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700018517.mm.1 TC1700001949.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0