Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600017328.mm.1 |
NM_001146689.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600017328.mm.1 |
NM_001146689.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600017328.mm.1 |
NM_007971.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600017328.mm.1 |
NM_007971.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600017328.mm.1 |
TC0600002200.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600017327.mm.1 |
NM_001146689.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600017327.mm.1 |
NM_001146689.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600017327.mm.1 |
NM_007971.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600017327.mm.1 |
NM_007971.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600017327.mm.1 |
TC0600002200.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600017326.mm.1 |
NM_001146689.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600017326.mm.1 |
NM_001146689.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600017326.mm.1 |
NM_007971.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600017326.mm.1 |
NM_007971.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600017326.mm.1 |
TC0600002200.mm.1 |
MTA TC |