This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01159240_g1
See other EZH2 GE Assays ›
Gene Symbol:
Ezh2
Gene Name:
enhancer of zeste homolog 2 (Drosophila)
Gene Aliases:
Enx-1, Enx1h, KMT6, mKIAA4065
Chromosome Location:
Chr.6: 47530274 - 47595340 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01159240_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ezh2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001146689.1 NP_001140161.1 13 - 14 1663 76
NM_007971.2 NP_031997.2 13 - 14 1675 76
GenBank mRNA AK157853.1 - 13 - 14 1676 76
AK220174.1 - 13 - 14 1630 76
BC003772.1 - 13 - 14 1584 3492110 76
BC016391.1 - 13 - 14 1597 3586689 76
BC079538.1 - 13 - 14 1614 6817366 76
U52951.1 - 13 - 14 1665 76

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ezh2

Panther Classification:

Molecular Function -

histone modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
regulation of protein phosphorylation
DNA methylation
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
regulation of gene expression
negative regulation of gene expression
positive regulation of epithelial to mesenchymal transition
regulation of gliogenesis
skeletal muscle satellite cell maintenance involved in skeletal muscle regeneration
chromatin modification
histone methylation
peptidyl-lysine methylation
cerebellar cortex development
methylation
negative regulation of transcription elongation from RNA polymerase II promoter
regulation of cell proliferation
regulation of circadian rhythm
positive regulation of MAP kinase activity
negative regulation of sequence-specific DNA binding transcription factor activity
positive regulation of GTPase activity
negative regulation of epidermal cell differentiation
negative regulation of gene expression, epigenetic
negative regulation of transcription, DNA-templated
negative regulation of retinoic acid receptor signaling pathway
rhythmic process
regulation of neurogenesis
negative regulation of striated muscle cell differentiation
cellular response to hydrogen peroxide
G1 to G0 transition
histone H3-K27 methylation
protein localization to chromatin
positive regulation of protein serine/threonine kinase activity
positive regulation of dendrite development
negative regulation of G1/S transition of mitotic cell cycle
regulatory region DNA binding
core promoter binding
chromatin binding
RNA binding
protein binding
methyltransferase activity
protein-lysine N-methyltransferase activity
transferase activity
histone-lysine N-methyltransferase activity
chromatin DNA binding
histone methyltransferase activity
sequence-specific DNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0