This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01158964_m1
See other FIGF GE Assays ›
Gene Symbol:
Figf
Gene Name:
c-fos induced growth factor
Gene Aliases:
AI325264, VEGF-D, Vegfd
Chromosome Location:
Chr.X: 164372998 - 164402650 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01158964_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Figf
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BC080770.1 - 2 - 3 568 30286444 69
D89628.1 - 2 - 3 400 69
X99572.1 - 2 - 3 587 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00001667.mm.1

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

growth factor

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00010667.mm.1 NM_010216.2 Refseq
Affymetrix Mouse Clariom S Array PSR0X00010667.mm.1 TC0X00001667.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0