This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01158962_m1
See other AP1S2 GE Assays ›
Gene Symbol:
Ap1s2
Gene Name:
adaptor-related protein complex 1, sigma 2 subunit
Gene Aliases:
1500012A13Rik, AI853648, EST1
Chromosome Location:
Chr.X: 163907377 - 163933666 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01158962_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ap1s2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001290378.1 NP_001277307.1 4 - 5 688 93
GenBank mRNA AK167690.1 - 4 - 5 563 93
AK211598.1 - 2 - 3 93

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

vesicle coat protein

Gene Ontology Categories:

Function(s) Process(es)

transport
intracellular protein transport
visual learning
protein transport
vesicle-mediated transport
synaptic vesicle recycling
fat cell differentiation
neuromuscular process controlling balance
adipose tissue development
protein binding
protein transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0