This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01158610_m1
See other SLC25A13 GE Assays ›
Gene Symbol:
Slc25a13
Gene Name:
solute carrier family 25 (mitochondrial carrier, adenine nucleotide translocator), member 13
Gene Aliases:
AI785475, Ctrn
Chromosome Location:
Chr.6: 6041218 - 6217233 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01158610_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc25a13
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001177572.1 NP_001171043.1 14 - 15 1611 78
NM_015829.3 NP_056644.1 14 - 15 1614 78
GenBank mRNA AF164632.1 - 14 - 15 1559 78
AK002829.1 - 14 - 15 1489 78
AK012670.1 - 14 - 15 1502 78
AK029020.1 - 14 - 15 1542 78
AK150496.1 - 14 - 15 1556 78
AK151180.1 - 14 - 15 1556 78
AK152237.1 - 14 - 15 1556 78
AK190917.1 - 1 - 2 72 78
BC016571.1 - 8 - 9 748 3600909 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600001813.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc25a13

Panther Classification:

Molecular Function -

transporter secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

ATP biosynthetic process
transport
aspartate transport
L-glutamate transport
malate-aspartate shuttle
cellular respiration
response to calcium ion
transmembrane transport
L-glutamate transmembrane transport
L-aspartate transmembrane transport
L-glutamate transmembrane transporter activity
calcium ion binding
L-aspartate transmembrane transporter activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600014383.mm.1 NM_001177572.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600014383.mm.1 NM_001177572.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600014383.mm.1 NM_015829.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600014383.mm.1 NM_015829.3 Refseq
Affymetrix Mouse Clariom S Array PSR0600014383.mm.1 TC0600001813.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0