This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01158441_m1
See other TCAF2 GE Assays ›
Gene Symbol:
Tcaf2
Gene Name:
TRPM8 channel-associated factor 2
Gene Aliases:
Fam115c
Chromosome Location:
Chr.6: 42623043 - 42645041 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01158441_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tcaf2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_146174.1 NP_666286.1 6 - 7 2350 90
GenBank mRNA AK131169.1 - 6 - 7 2563 90
BC008555.1 - 6 - 7 2367 3592166 90
BC011487.1 - 6 - 7 2350 3499593 90

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600002177.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tcaf2

Gene Ontology Categories:

Function(s) Process(es)

hematopoietic progenitor cell differentiation
transport
negative regulation of anion channel activity
positive regulation of cell migration
positive regulation of protein targeting to membrane
ion channel binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600017156.mm.1 NM_146174.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600017156.mm.1 NM_146174.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600017156.mm.1 TC0600002177.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600017155.mm.1 NM_146174.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600017155.mm.1 NM_146174.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600017155.mm.1 TC0600002177.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0