This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01158403_m1
See other FAM71F1 GE Assays ›
Gene Symbol:
Fam71f1
Gene Name:
family with sequence similarity 71, member F1
Gene Aliases:
GARI-L1, Gm764, NYD-SP18
Chromosome Location:
Chr.6: 29313482 - 29336022 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01158403_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fam71f1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001289663.1 NP_001276592.1 1 - 2 307 89
NM_001289664.1 NP_001276593.1 1 - 2 307 89
NM_207258.3 NP_997141.2 1 - 2 307 89
GenBank mRNA AB473725.1 - 1 - 2 248 89
AK133908.1 - 1 - 2 308 89
BC048651.1 - 1 - 2 308 89
BC147513.1 - 1 - 2 290 9056505 89
BC172074.1 - 1 - 2 290 9054360 89
BC172086.1 - 1 - 2 290 9054372 89

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600000228.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Fam71f1

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600001451.mm.1 NM_001289663.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600001451.mm.1 NM_001289664.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600001451.mm.1 NM_001289665.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600001451.mm.1 NM_207258.3 Refseq
Affymetrix Mouse Clariom S Array PSR0600001451.mm.1 TC0600000228.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600001453.mm.1 NM_001289663.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600001453.mm.1 NM_001289664.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600001453.mm.1 NM_001289665.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600001453.mm.1 NM_207258.3 Refseq
Affymetrix Mouse Clariom S Array PSR0600001453.mm.1 TC0600000228.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600001450.mm.1 NM_001289663.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600001450.mm.1 NM_001289664.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600001450.mm.1 NM_001289665.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600001450.mm.1 NM_207258.3 Refseq
Affymetrix Mouse Clariom S Array PSR0600001450.mm.1 TC0600000228.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm