This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01157033_g1
See other XPOT GE Assays ›
Gene Symbol:
Xpot
Gene Name:
exportin, tRNA (nuclear export receptor for tRNAs)
Gene Aliases:
1110004L07Rik, 3110065H13Rik, AI452076, C79645, EXPORTIN-T
Chromosome Location:
Chr.10: 121587380 - 121626316 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01157033_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Xpot
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081056.1 NP_001074525.1 15 - 16 1946 109
GenBank mRNA AK029113.1 - 10 - 11 1399 109

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

RNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

tRNA export from nucleus
transport
intracellular protein transport
tRNA binding
RNA binding
Ran GTPase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000022742.mm.1 NM_001081056.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000022742.mm.1 NM_001081056.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000022742.mm.1 TC1000003050.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0