This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01151324_m1
See other PRKCSH GE Assays ›
Gene Symbol:
Prkcsh
Gene Name:
protein kinase C substrate 80K-H
Gene Aliases:
80K-H, PKCSH
Chromosome Location:
Chr.9: 22002988 - 22014245 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01151324_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Prkcsh
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001293650.1 NP_001280579.1 5 - 6 508 86
NM_001293651.1 NP_001280580.1 5 - 6 512 86
NM_008925.2 NP_032951.1 5 - 6 508 86
GenBank mRNA AK137661.1 - 5 - 6 475 86
AK153931.1 - 5 - 6 483 86
AK159705.1 - 5 - 6 479 86
BC009816.1 - 5 - 6 441 4008182 86
CD807078.1 - 5 - 6 477 86
U92794.1 - 5 - 6 461 86

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900000210.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Prkcsh

Panther Classification:

Molecular Function -

protein-binding activity modulator

Gene Ontology Categories:

Function(s) Process(es)

in utero embryonic development
liver development
N-glycan processing
nitrogen compound metabolic process
negative regulation of neuron projection development
protein heterooligomerization
renal system development
RNA binding
protein kinase C binding
calcium ion binding
protein binding
ion channel binding
metal ion binding
phosphoprotein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900001600.mm.1 NM_008925.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900001600.mm.1 TC0900000210.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0