Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000006938.mm.1 |
NM_001285853.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000006938.mm.1 |
NM_001285854.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000006938.mm.1 |
NM_010768.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000006938.mm.1 |
TC1000000889.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000006936.mm.1 |
NM_001285853.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000006936.mm.1 |
NM_001285854.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000006936.mm.1 |
NM_010768.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000006936.mm.1 |
TC1000000889.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000006937.mm.1 |
NM_001285853.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000006937.mm.1 |
NM_001285854.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000006937.mm.1 |
NM_010768.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000006937.mm.1 |
TC1000000889.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000006939.mm.1 |
NM_001285853.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000006939.mm.1 |
NM_001285854.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000006939.mm.1 |
NM_010768.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000006939.mm.1 |
TC1000000889.mm.1 |
MTA TC |