This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01151038_g1
See other MRPL4 GE Assays ›
Gene Symbol:
Mrpl4
Gene Name:
mitochondrial ribosomal protein L4
Gene Aliases:
1110017G11Rik
Chromosome Location:
Chr.9: 21002605 - 21008839 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01151038_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mrpl4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_023167.2 NP_075656.2 6 - 7 757 76
GenBank mRNA AB041599.1 - 5 - 6 509 76
AB049634.1 - 6 - 7 756 76
AK002464.1 - 6 - 7 593 76
AK003746.2 - 6 - 7 714 76
BC037064.1 - 6 - 7 590 76
BC039983.1 - 6 - 7 725 4952757 76
BC051685.1 - 6 - 7 570 76
BC061095.1 - 6 - 7 573 6588836 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900000183.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mrpl4

Panther Classification:

Molecular Function -

ribosomal protein

Gene Ontology Categories:

Function(s) Process(es)

translation
RNA binding
structural constituent of ribosome
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900001199.mm.1 NM_023167.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900001199.mm.1 NM_023167.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900001199.mm.1 TC0900000183.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-c694fbbc8-74xkl:80/100.66.128.6:80.
git-commit: 2cf2d5f40d96ab2bfa499d940143ca22e465fdb8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0