This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01151033_g1
See other MRPL4 GE Assays ›
Gene Symbol:
Mrpl4
Gene Name:
mitochondrial ribosomal protein L4
Gene Aliases:
1110017G11Rik
Chromosome Location:
Chr.9: 21002605 - 21008839 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01151033_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mrpl4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_023167.2 NP_075656.2 1 - 2 256 54
GenBank mRNA AB049634.1 - 1 - 2 255 54
AK002464.1 - 1 - 2 92 54
AK003746.2 - 1 - 2 214 54
BC023078.1 - 1 - 2 37 54
BC037064.1 - 1 - 2 89 54
BC039983.1 - 1 - 2 224 4952757 54
BC051685.1 - 1 - 2 69 54
BC061095.1 - 1 - 2 72 6588836 54

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900000183.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mrpl4

Panther Classification:

Molecular Function -

ribosomal protein

Gene Ontology Categories:

Function(s) Process(es)

translation
RNA binding
structural constituent of ribosome
poly(A) RNA binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900001190.mm.1 NM_023167.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900001190.mm.1 NM_023167.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900001190.mm.1 TC0900000183.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900001188.mm.1 NM_023167.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900001188.mm.1 NM_023167.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900001188.mm.1 TC0900000183.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900001192.mm.1 NM_023167.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900001192.mm.1 NM_023167.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900001192.mm.1 TC0900000183.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900001191.mm.1 NM_023167.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900001191.mm.1 NM_023167.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900001191.mm.1 TC0900000183.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900001189.mm.1 NM_023167.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900001189.mm.1 NM_023167.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900001189.mm.1 TC0900000183.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 744893 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0