This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01150302_m1
See other ADORA2A GE Assays ›
Gene Symbol:
Adora2a
Gene Name:
adenosine A2a receptor
Gene Aliases:
A2AAR, A2aR, AA2AR
Chromosome Location:
Chr.10: 75316860 - 75334792 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01150302_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Adora2a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009630.3 NP_033760.2 2 - 3 839 65
GenBank mRNA AK037787.1 - 1 - 2 61 65
AK206618.1 - 1 - 2 228 65
BC110692.1 - 1 - 2 339 30242398 65
CX241487.1 - 2 - 3 645 65
U05672.1 - 1 - 2 563 65

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Adora2a

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

synaptic transmission, dopaminergic
adenosine receptor signaling pathway
response to amphetamine
regulation of transcription, DNA-templated
negative regulation of protein kinase activity
signal transduction
G-protein coupled receptor signaling pathway
adenylate cyclase-activating G-protein coupled receptor signaling pathway
protein kinase C-activating G-protein coupled receptor signaling pathway
synaptic transmission, cholinergic
locomotory behavior
negative regulation of cell proliferation
positive regulation of adenylate cyclase activity involved in G-protein coupled receptor signaling pathway
positive regulation of glutamate secretion
positive regulation of acetylcholine secretion, neurotransmission
regulation of norepinephrine secretion
positive regulation of cAMP biosynthetic process
response to caffeine
positive regulation of synaptic transmission, GABAergic
synaptic transmission, glutamatergic
positive regulation of urine volume
positive regulation of renal sodium excretion
negative regulation of locomotion
vasodilation
eating behavior
negative regulation of vascular permeability
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
response to alkaloid
negative regulation of neuron apoptotic process
positive regulation of circadian sleep/wake cycle, sleep
negative regulation of alpha-beta T cell activation
astrocyte activation
regulation of synaptic plasticity
neuron projection morphogenesis
positive regulation of protein secretion
negative regulation of inflammatory response
regulation of mitochondrial membrane potential
membrane depolarization
regulation of calcium ion transport
positive regulation of synaptic transmission, glutamatergic
excitatory postsynaptic potential
inhibitory postsynaptic potential
prepulse inhibition
positive regulation of apoptotic signaling pathway
G-protein coupled adenosine receptor activity
signal transducer activity
G-protein coupled receptor activity
enzyme binding
type 5 metabotropic glutamate receptor binding
identical protein binding
protein heterodimerization activity
alpha-actinin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0