This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01150068_g1
See other DOS GE Assays ›
Gene Symbol:
Dos
Gene Name:
downstream of Stk11
Gene Aliases:
9630008F14, R29144/1
Chromosome Location:
Chr.10: 80130434 - 80140136 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01150068_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dos
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001195268.1 NP_001182197.1 5 - 6 623 117
NR_036582.1 7 - 8 959 117
GenBank mRNA AK137009.1 - 7 - 8 959 117
BC052075.1 - 6 - 7 677 117
BC078442.1 - 1 - 2 178 117

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000002491.mm.1

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

voltage-gated ion channel

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000018361.mm.1 NM_001195268.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000018361.mm.1 NM_001195268.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000018361.mm.1 NR_036582.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000018361.mm.1 NR_036582.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000018361.mm.1 TC1000002491.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0