This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01149687_m1
See other AIRE GE Assays ›
Gene Symbol:
Aire
Gene Name:
autoimmune regulator (autoimmune polyendocrinopathy candidiasis ectodermal dystrophy)
Gene Aliases:
-
Chromosome Location:
Chr.10: 78030022 - 78043610 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01149687_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Aire
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001271556.1 NP_001258485.1 9 - 10 1194 116
NM_001271557.1 NP_001258486.1 9 - 10 1191 116
NM_001271558.1 NP_001258487.1 9 - 10 1182 116
NM_001271559.1 NP_001258488.1 9 - 10 1179 116
GenBank mRNA AF128122.1 - 9 - 10 1164 116
AF128123.1 - 9 - 10 1161 116
AF128124.1 - 9 - 10 1152 116
AF128125.1 - 9 - 10 1149 116
AK131938.1 - 2 - 3 493 116

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
humoral immune response
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II regulatory region sequence-specific DNA binding
DNA binding
chromatin binding
zinc ion binding
histone binding
identical protein binding
transcription regulatory region DNA binding
translation regulator activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0