This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01149682_g1
See other AIRE GE Assays ›
Gene Symbol:
Aire
Gene Name:
autoimmune regulator (autoimmune polyendocrinopathy candidiasis ectodermal dystrophy)
Gene Aliases:
-
Chromosome Location:
Chr.10: 78030022 - 78043610 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01149682_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Aire
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001271549.1 NP_001258478.1 2 - 3 397 91
NM_001271550.1 NP_001258479.1 2 - 3 397 91
NM_001271551.1 NP_001258480.1 2 - 3 397 91
NM_001271552.1 NP_001258481.1 2 - 3 397 91
NM_001271553.1 NP_001258482.1 2 - 3 397 91
NM_001271554.1 NP_001258483.1 2 - 3 397 91
NM_001271555.1 NP_001258484.1 2 - 3 397 91
NM_001271556.1 NP_001258485.1 2 - 3 397 91
NM_001271557.1 NP_001258486.1 2 - 3 397 91
NM_001271558.1 NP_001258487.1 2 - 3 397 91
NM_001271559.1 NP_001258488.1 2 - 3 397 91
NM_009646.2 NP_033776.1 2 - 3 397 91
NR_073358.1 2 - 3 397 91
GenBank mRNA AF079536.1 - 2 - 3 367 91
AF128115.1 - 2 - 3 367 91
AF128116.1 - 2 - 3 367 91
AF128117.1 - 2 - 3 367 91
AF128118.1 - 2 - 3 367 91
AF128119.1 - 2 - 3 367 91
AF128120.1 - 2 - 3 367 91
AF128121.1 - 2 - 3 367 91
AF128122.1 - 2 - 3 367 91
AF128123.1 - 2 - 3 367 91
AF128124.1 - 2 - 3 367 91
AF128125.1 - 2 - 3 367 91
AF128772.1 - 2 - 3 310 91
AF128773.1 - 2 - 3 310 91
AJ132243.1 - 2 - 3 310 91
AJ243821.1 - 2 - 3 310 91
AK153690.1 - 3 - 4 451 91
BC103510.1 - 2 - 3 359 40039055 91
BC103511.1 - 2 - 3 359 40039054 91
BC103518.1 - 2 - 3 359 40039058 91
EU625343.3 - 2 - 3 327 91
JF267399.1 - 2 - 3 327 91

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000002440.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Aire

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
humoral immune response
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II regulatory region sequence-specific DNA binding
DNA binding
chromatin binding
zinc ion binding
histone binding
identical protein binding
transcription regulatory region DNA binding
translation regulator activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0