This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01149115_g1
See other GCLC GE Assays ›
Gene Symbol:
Gclc
Gene Name:
glutamate-cysteine ligase, catalytic subunit
Gene Aliases:
D9Wsu168e, GLCL-H, Ggcs-hs, Glclc
Chromosome Location:
Chr.9: 77754516 - 77794489 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01149115_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gclc
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010295.2 NP_034425.1 9 - 10 1360 75
GenBank mRNA AK144339.1 - 9 - 10 1361 75
AK160876.1 - 9 - 10 1120 75
AK169676.1 - 9 - 10 1333 75
BC019374.1 - 9 - 10 1117 4195425 75
U85414.1 - 9 - 10 1085 75
U85498.1 - 9 - 10 1085 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900001038.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gclc

Panther Classification:

Molecular Function -

ligase

Gene Ontology Categories:

Function(s) Process(es)

cysteine metabolic process
glutamate metabolic process
glutathione metabolic process
glutathione biosynthetic process
response to oxidative stress
apoptotic mitochondrial changes
response to heat
response to xenobiotic stimulus
response to hormone
L-ascorbic acid metabolic process
negative regulation of protein ubiquitination
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
negative regulation of apoptotic process
negative regulation of neuron apoptotic process
cell redox homeostasis
negative regulation of transcription, DNA-templated
response to arsenic-containing substance
regulation of blood vessel size
response to nitrosative stress
regulation of mitochondrial depolarization
negative regulation of extrinsic apoptotic signaling pathway
nucleotide binding
magnesium ion binding
glutamate-cysteine ligase activity
ATP binding
glutamate binding
ligase activity
ADP binding
protein heterodimerization activity
coenzyme binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900008137.mm.1 NM_010295.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900008137.mm.1 NM_010295.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900008137.mm.1 TC0900001038.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0