This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01149064_g1
See other SPPL2B GE Assays ›
Gene Symbol:
Sppl2b
Gene Name:
signal peptide peptidase like 2B
Gene Aliases:
3110056O03Rik, AW550292, PSL1
Chromosome Location:
Chr.10: 80855248 - 80868708 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01149064_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sppl2b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_175195.3 NP_780404.2 7 - 8 882 65
GenBank mRNA AK046804.1 - 7 - 8 901 65
AK148109.1 - 7 - 8 849 65
AK157906.1 - 7 - 8 827 65
AK165324.1 - 7 - 8 1007 65
AK166364.1 - 7 - 8 833 65
AK170375.1 - 7 - 8 848 65
BC034206.1 - 4 - 5 437 65
BC052094.1 - 5 - 6 556 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000000874.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sppl2b

Panther Classification:

Molecular Function -

aspartic protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
membrane protein ectodomain proteolysis
membrane protein intracellular domain proteolysis
regulation of immune response
aspartic-type endopeptidase activity
peptidase activity
hydrolase activity
aspartic endopeptidase activity, intramembrane cleaving
protein homodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000006719.mm.1 NM_175195.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000006719.mm.1 NM_175195.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000006719.mm.1 TC1000000874.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0