This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01147321_g1
See other SLTM GE Assays ›
Gene Symbol:
Sltm
Gene Name:
SAFB-like, transcription modulator
Gene Aliases:
5730455C01Rik, 5730555F13Rik, 9130215G10Rik, Met
Chromosome Location:
Chr.9: 70541796 - 70592237 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01147321_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sltm
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025690.3 NP_079966.2 10 - 11 1433 128
NM_026337.1 NP_080613.1 10 - 11 1471 128
GenBank mRNA AF462146.1 - 10 - 11 1433 128
AK011815.1 - 10 - 11 1419 128
AK017839.1 - 10 - 11 1332 128
AK032003.1 - 2 - 3 1589 128
AK045723.1 - 4 - 5 533 128
AK077578.1 - 10 - 11 1494 128
AK082294.1 - 7 - 8 2349 128
AK134756.1 - 1 - 2 112 128
BC139202.1 - 10 - 11 1423 8862224 128
BC145577.1 - 10 - 11 1423 128

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900000938.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sltm

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
apoptotic process
biological_process
nucleotide binding
nucleic acid binding
RNA binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900007236.mm.1 NM_025690.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900007236.mm.1 NM_025690.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900007236.mm.1 NM_026337.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900007236.mm.1 NM_026337.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900007236.mm.1 TC0900000938.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0