This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01147243_g1
See other ATG4D GE Assays ›
Gene Symbol:
Atg4d
Gene Name:
autophagy related 4D, cysteine peptidase
Gene Aliases:
9830134P12Rik, APG4-D, Apg4d, Atg4dl, Autl4
Chromosome Location:
Chr.9: 21265285 - 21274837 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01147243_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atg4d
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_153583.10 NP_705811.8 7 - 8 1238 68
GenBank mRNA AK036282.1 - 7 - 8 1226 68
AK036571.1 - 7 - 8 1227 68
AK081002.1 - 7 - 8 1225 68
AK081605.1 - 7 - 8 1216 68
AK087258.1 - 7 - 8 1430 68
AK154130.1 - 7 - 8 1231 68
BC030861.1 - 7 - 8 1215 4208700 68
BC069851.1 - 7 - 8 1190 6490007 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900000191.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Atg4d

Panther Classification:

Molecular Function -

cysteine protease

Gene Ontology Categories:

Function(s) Process(es)

autophagosome assembly
mitophagy
C-terminal protein lipidation
proteolysis
protein targeting to membrane
transport
autophagy
apoptotic process
cellular response to nitrogen starvation
protein transport
protein processing
nucleophagy
protein delipidation
cysteine-type endopeptidase activity
peptidase activity
cysteine-type peptidase activity
hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900001285.mm.1 NM_153583.10 Refseq
Affymetrix Mouse Clariom S Array PSR0900001285.mm.1 TC0900000191.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0