This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01147189_g1
See other ZFR2 GE Assays ›
Gene Symbol:
Zfr2
Gene Name:
zinc finger RNA binding protein 2
Gene Aliases:
2010013I23Rik, 9130206N08Rik, AW045600, mKIAA1086
Chromosome Location:
Chr.10: 81233163 - 81252123 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01147189_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Zfr2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001034895.3 NP_001030067.2 11 - 12 1755 72
GenBank mRNA AK033649.1 - 4 - 5 504 72
AK122438.1 - 4 - 5 3363 72
BC072571.1 - 10 - 11 2424 72
BF449481.1 - 1 - 2 130 72

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000000888.mm.1

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

RNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000006921.mm.1 NM_001034895.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000006921.mm.1 NM_001034895.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000006921.mm.1 TC1000000888.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0