This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01147139_m1
See other ZFP426 GE Assays ›
Gene Symbol:
Zfp426
Gene Name:
zinc finger protein 426
Gene Aliases:
2900057C04Rik, KRAB1, Zfo61, Zfp68-rs1, Znf426, mKIAA4237
Chromosome Location:
Chr.9: 20468548 - 20493922 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01147139_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Zfp426
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001110309.1 NP_001103779.1 6 - 7 554 154
NM_146221.4 NP_666333.1 4 - 5 692 154
GenBank mRNA AK036912.1 - 6 - 7 446 154
AK085144.1 - 4 - 5 693 154
AK220563.1 - 4 - 5 1394 154
BC024805.1 - 6 - 7 550 5359610 154

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Zfp426

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
biological_process
nucleic acid binding
DNA binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0